Learn More
Brady™ Water Dissolvable Paper Tape
These water dissolvable paper labels dissolve completely in water, leaving no residue behind.
Brand: Brady™ 149608
This item is not returnable.
View return policy
Description
These water dissolvable paper labels are self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to, and without leaving any residue.
- Reduces labor required to remove labels
- Reduces maintenance to filters and drains
- Enables easy reidentification of reusable items in a range of industries
- Dissolves completely in water, leaving no adhesive residue behind
Specifications
Black on White | |
Paper | |
0.75 in. | |
19.1 mm | |
0.08 mm | |
Matte |
BMP21 Label Printer, BMP21-PLUS Label Printer, BMP21-LAB Label Printer | |
21 ft. | |
6.4 m | |
0.003 in. | |
Acrylic |