Learn More
Brady™ Water Dissolvable Paper Tape
These water dissolvable paper labels dissolve completely in water, leaving no residue behind.
Brand:  Brady™ 149608
Product Code. 17374101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
These water dissolvable paper labels are self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to, and without leaving any residue.
  • Reduces labor required to remove labels
  • Reduces maintenance to filters and drains
  • Enables easy reidentification of reusable items in a range of industries
  • Dissolves completely in water, leaving no adhesive residue behind
Specifications
Specifications
Black on White
Paper
0.75 in.
19.1 mm
0.08 mm
Matte
BMP21 Label Printer, BMP21-PLUS Label Printer, BMP21-LAB Label Printer
21 ft.
6.4 m
0.003 in.
Acrylic