missing translation for 'onlineSavingsMsg'
Learn More

Rogo Sampaic™ Watch Glasses

Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.

Brand:  Rogo Sampaic™ KBN019

 View more versions of this product

Product Code. 11724146

  • 123.00€ / Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

3 L
Watch Glass
500 °C
Borosilicate Glass
Beaker
160 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.