Learn More
Rogo Sampaic™ Watch Glasses
Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.
Brand:  Rogo Sampaic™ KBN013
 View more versions of this product
Product Code. 11702724
  • 110.00€ / Pack of 20
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Autoclavable
Borosilicate Glass
Beaker
60 mm
100 mL
Watch Glass
500 °C