Learn More
Vitlab™ Wash Bottle Cap
Click to view available options
Material:
Polypropylene
Description
- Screw cap and spray tube with drawn-out tip, made of PP
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
Specifications
Specifications
Includes | Spray tube with drawn-out tip |
Material | Polypropylene |
Thread Size (Metric) | GL 18 |
Closure Type | Screw Cap |