missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Warning Sign, wear protective clothing

Use this sign to help increase laboratory safety. Brady™ Warning Signs feature adhesive polyethylene and text-free images to prevent misunderstanding.

Brand:  Brady™ 806870

Product Code. 11997381

  • 4.45€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Diameter: 100mm
  • Material: Adhesive PE
Specifications

Specifications

PIC 2209-DIA 100-B7541 FF
Polyethylene
Wear lab coat
Polyethylene
100 mm
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.