missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0238429
Selective positive allosteric modulator of M5 receptors
162.00€ - 636.00€
Specifications
Purity | 0.97 |
---|---|
Content And Storage | Store at -20°C |
Product Type | VU 0238429 |
Molecular Weight (g/mol) | 351.28 |
Inhibitors | VU 0238429 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18043932
|
Tocris Bioscience™
3634/10 |
10 mg |
162.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18093852
|
Tocris Bioscience™
3634/50 |
50 mg |
636.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective positive allosteric modulator of M5 receptors (EC50 values are 1.16, >30 and >30 μM at M5, M1 and M3 receptors respectively). Displays no activity at M2 and M4 receptors.Specifications
0.97 | |
VU 0238429 | |
VU 0238429 |
Store at -20°C | |
351.28 |
For Research Use Only