Learn More
Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™ is thick-walled, clearly marked and labeled, and has a solid finish.
Brand:  Karl Hecht™ 42097084
 View more versions of this product
Product Code. 10516162
  • 51.00€ / Each
Explore more special offers
Description
Description
  • Duran™ borosilicate glass 3.3.
  • Fused blue graduation.
  • Standard ground without stopper.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
DIN EN ISO 1042
Borosilicate Glass (3.3)
Volumetric Flask
NS 29
1000 mL
Wide