Learn More
Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™ is thick-walled, clearly marked and labeled, and has a solid finish.
Brand: Karl Hecht™ 42097080
This item is not returnable.
View return policy
Description
- Duran™ borosilicate glass 3.3.
- Fused blue graduation.
- Standard ground without stopper.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
DIN EN ISO 1042 | |
Borosilicate Glass (3.3) | |
Volumetric Flask | |
NS 24 |
1000 mL | |
Standard | |
Yes |