missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methylene chloride
missing translation for 'unitSize'
Each
Pack of 12
Pack of 6
Description
- Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
- VENT-CAP practically eliminates leakage and prevents almost all static overpressure
- Color-coded for easy visual identification of the media
- Material name in German, English, French and Spanish
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Saving and storing |
| Includes | Spray tube and imprint |
| Material | LDPE |
| No. per Pack | 12 Pack |
| Closure Color | Red |
| Closure Included | Yes |
| Mouth | Narrow |
| Height (Metric) | 180 mm |
| Product Type | Wash Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction