Learn More
VistaLab™ Technologies Reagent Reservoirs
Unique trough-within-trough design maximizes reagent recovery
Specifications
Certifications/Compliance | Pyrogen-free |
---|---|
Type | Reagent Reservoir |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Quantity | Price | Quantity & Availability | ||
11915732
|
VistaLab™ Technologies
3054-1000 |
100 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11925732
|
VistaLab™ Technologies
3054-1001 |
100 mL | Sterile | Individually Wrapped | 50/Cs. |
N/A
|
N/A | ||
11945732
|
VistaLab™ Technologies
3054-1004 |
25 mL (divided) | Sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A
|
||
11925742
|
VistaLab™ Technologies
3054-3001 |
100 mL (with lid) | Sterile | Individually Wrapped | 50/Cs. |
N/A
|
N/A | ||
11955732
|
VistaLab™ Technologies
3054-1003 |
25 mL | Sterile | Individually Wrapped | 50/Cs. |
N/A
|
N/A | ||
11975732
|
VistaLab™ Technologies
3054-1006 |
50 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11985732
|
VistaLab™ Technologies
3054-1007 |
50 mL | Sterile | Individually Wrapped | 50/Cs. |
N/A
|
N/A | ||
11935722
|
VistaLab™ Technologies
3054-2001 |
100 mL | Sterile | Individually Wrapped | 100/Cs. |
N/A
|
N/A | ||
11995732
|
VistaLab™ Technologies
3054-2000 |
100 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
N/A
|
N/A | ||
11965722
|
VistaLab™ Technologies
3054-2006 |
50 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
N/A
|
N/A | ||
11945722
|
VistaLab™ Technologies
3054-2002 |
25 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
N/A
|
N/A | ||
11905742
|
VistaLab™ Technologies
3054-2007 |
50 mL | Sterile | Individually Wrapped | 100/Cs. |
N/A
|
N/A | ||
11955722
|
VistaLab™ Technologies
3054-2003 |
25 mL | Sterile | Individually Wrapped | 100/Cs. |
N/A
|
N/A | ||
11975742
|
VistaLab™ Technologies
4054-1002 |
25 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11985742
|
VistaLab™ Technologies
4054-1004 |
25 mL (divided) | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11915752
|
VistaLab™ Technologies
4054-1006 |
50 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11965742
|
VistaLab™ Technologies
4054-1000 |
100 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11935732
|
VistaLab™ Technologies
3054-1002 |
25 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
N/A
|
N/A | ||
11965732
|
VistaLab™ Technologies
3054-1005 |
25 mL (divided) | Sterile | Individually Wrapped | 50/Cs. |
N/A
|
N/A
|
||
15742962
|
VistaLab™ Technologies
3054-1012 |
10 mL | Sterile | Bulk | 100/Cs. |
N/A
|
N/A | ||
16445094
|
VistaLab™ Technologies
3054-1013 |
10 mL | Sterile | Individually packed | 50/Cs. |
N/A
|
N/A | ||
16455094
|
VistaLab™ Technologies
3054-2012 |
10 mL | Sterile | Bulk | 200 in. 20 bags of 10 |
N/A
|
N/A | ||
16465094
|
VistaLab™ Technologies
3054-2013 |
10 mL | Sterile | Individually packed | 100/Cs. |
N/A
|
N/A | ||
16405104
|
VistaLab™ Technologies
4054-1012 |
10 mL | Non-sterile | Bulk | 100/Cs. |
N/A
|
N/A | ||
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
- 100mL model (Cat. No. 21-385-111) is supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Order Info
Reservoir lid sold separately (Cat. No. 21-385-110).
Specifications
Pyrogen-free | |
Reagent Reservoir | |
Single-use |