missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66305-1-IG-150UL
This item is not returnable.
View return policy
Description
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.Specifications
Vinculin | |
Monoclonal | |
0.78 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18206, P26234, P85972, Q64727 | |
VCL | |
Vinculin Fusion Protein Ag24946 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B5A7 | |
Unconjugated | |
VCL | |
CMD1W, Metavinculin, MVCL, VCL, Vinculin | |
Mouse | |
Protein G | |
RUO | |
22330, 305679, 396974, 7414 | |
-20°C | |
Liquid |