missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Vildagliptin anti-diabetic agent
Vildagliptin is a potent inhibitor of dipeptidyl peptidase IV (DPP IV), an enzyme that cleaves glucagon-like peptide 1 (GLP-1) (1).
Brand: Novus Biologicals™ NBP2-62759-20MG
This item is not returnable.
View return policy
Description
PubChem ID: 11077541
Application Notes: May be dissolved in DMSO (45 mg/mL); or DMF (20 mg/mL); or ethanol (20 mg/mL).

Specifications
274901-16-5 | |
C17H25N3O2 | |
20 mg | |
Vildagliptin anti-diabetic agent |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
>98% (HPLC); NMR (conforms) | |
Synthetic | |
Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Vildagliptin anti-diabetic agent > 20 mg
Spot an opportunity for improvement?Share a Content Correction