Learn More
Tocris Bioscience™ VH 032 amide-alkylC5-acid
VHL ligand with alkyl linker and terminal acid for onward chemistry
Brand:  Tocris Bioscience™
213.37 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18014792
  • 264.00€ / 25mg
Explore more special offers
Description
Description
VH 032 amide-alkylC5-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC™ research and development; incorporates an E3 ligase ligand plus alkylC5 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
7-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-7-oxoheptanoic acid
25 mg
C29H40N4O6S
2162120-87-6
Degrader Building Blocks
95%