Learn More
Unity Lab Services™ Verification Report Dry Bath
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand: Unity Lab Services™ UMV-P932P
Additional Details : Weight : 0.01000kg
Description
- Checks on equipment specifications including: stability, homogeneity, setpoint deviation and indication if existing
- Measurements made simultaneously at five points on the block (one in the centre and at each of the four corners)
- Delivery carried out with reference equipment conforming to national standards
Specifications
Dry Baths | |
Verification Report |