Learn More
Emma Safety Footwear Vera XD Safety Shoes
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and direct resistant.
Brand:  Emma Safety Footwear MM50756444
 View more versions of this product
Product Code. 15928641
  • 111.00€ / 1 pair
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
Vera XD
Microfiber
44