Learn More
Veolia Water Systems™ UV Lamp
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
Brand:  Veolia Water Systems™ LC105
 View more versions of this product
Product Code. 11954897
  • 462.00€ / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
Specifications
Break down and photo-oxidize organic contaminants to polar or ionized species
Biopure 60/120, Biopure 7/15, Maxima MK1/MK2, Medica 30/60/100 MK2 & 3, Medica 7/15 MK1, Medica Pro, Medica-D 7/15, Medica-R 30/60/100, Medica-R 7/15, Medica-S 7/15, Optima 15/30/60 & Plus, Option 1/2/3/4/5/6, PureLab Elect 5/10, PureLab Option 7/15 And Ps, PureLab Option-E 25/50/80 (Elect), PureLab Option-E 5/10, PureLab Option-R 30/60, PureLab Option-R 7/15, PureLab Option-S 30/60, PureLab Ultra MK1, PureLab Ultra MK2