Learn More
Elga™ Pre-Treatment Cartridge
Protects RO membranes by removing particulates and chlorine.
Brand: Elga™ LC140
Description
- Pretreatment cartridge
- Helps to produce general laboratory grade water at better than single distilled quality
- Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets
Specifications
Glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets | |
Purelab™ Option-S series water purification systems |