missing translation for 'onlineSavingsMsg'
Learn More

MMM Medcenter™ Venticell Comfort Line Oven

For applications in biological research, production and quality control.

Brand:  MMM Medcenter™ MC001818

Product Code. 11461138

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Working temperature of from +10°C over ambient temperature up to 250°C/300°C
  • Patented system of forced air flow in the chamber ensures a homogenous temperature profile for all processes of drying, heating and sterilizing materials in laboratories
  • High operation comfort, precise temperature regulation and short times of temperature recovery in the chamber after the door opening
  • Suitable especially for materials of high humidity, for demanding and accurate tests and processes
  • Comfort model with a Fuzzy-Logic microprocessor control system
  • Control elements situated on a foil keyboard
  • Process information indicated on a LCD display
  • Output for a printer or PC connection
  • Validation possibility (IQ , OQ)
Specifications

Specifications

404 L
Venticell Comfort Line Oven
540 x 540 x 1410 mm
760 mm
790 mm
Ambient +10° to 250°C/300°C
540 x 540 x 1410 mm
790 mm
1910 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers