Learn More
Sonics Materials VCX-130PB Ultrasonic Processor
Product Code. 11763729
Click to view available options
Wattage:
130 W
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 11763729
Brand: Sonics Materials VCX130PB220
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Robust and reliable 130 watt Ultrasonic Processor with thumb-actuated pulser for small volume applications.
  • Ultrasonic processors for small volume applications
  • 130 watt ultrasonic processor with thumb-actuated pulser or footswitch
  • Safely process a wide range of organic and inorganic materials, from 150 microliters to 150 milliliters
  • Typical applications include biotechnology and pharmaceutical processing, including mixing, dispersion, and sample prep uses
  • Energy monitor, digital wattmeter, automatic tuning
  • Variable power output control
Specifications
Product Type Ultrasonic Processor
Wattage 130 W
Includes Thumb-actuated pulser