missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Semi-Rigid Tubing, UNF(M)
VapLock™ Adapters for Semi-Rigid Tubing, UNF(M) are designed for secure liquid-tight connections
59.75€
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | VapLock Solvent Containment Systems |
Product Code | Brand | Size | Thread Size (English) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Thread Size (English) | Material | Price | Quantity & Availability | |||
17182435
|
Cole-Parmer™
12020-03 |
1/16 in. OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Green / Blue PP / ETFE |
59.75€
Pack of 10 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
Description
- Color-coded for easy identification
- Features two-piece nut and ferrule system
Warranty
None
Specifications
Solvent Safety | |
VapLock Solvent Containment Systems |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title