Learn More
IKA™ UTL 25 Digital Inline ULTRA-TURRAX™ with UK Plug
UTL 25 digital Inline ULTRA-TURRAX™ for circulation or flow-through processing in the laboratory.
Brand:  IKA™ 0010006858
Product Code. 17593596
  • 11845.00€ / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Simple, compact and sturdy modular design
  • Sterilizable, autoclave-compatible
  • Table-top or stand-supported device, low space requirement
  • Easy disassembly
  • Large delivery capacity of 4.4 to 11.6 L/min. with open outlet (the mounting of a valve can reduce the flow rate)
  • For air-free, sterile, and inline suspension, emulsifying and deagglomeration
  • For vacuum or pressurized operation (up to 6 bar)
  • If the DK 25.11 is used, air induction is also prevented in batch operation
  • Not self-priming
  • A pump can be integrated between intake nozzle and vessel. As a result, viscous fluids can be processed
  • Not suitable for continuous operation or cyclical continuous operation
  • The hoses used should have an inner diameter size of 13 mm (inlet) and 9 mm (outlet) respectively to fit the DK 25.11 flow chamber
Specifications
Specifications
3000 to 25,000 rpm
50/60 Hz
13 mm
175 mm
73 dB
3.8 kg
230 V
26 mL
450 x 120 x 100 mm
Circulation or Flow-Through Processing
UK