Learn More
Testo™ USB Connection Cable
Connect enabled instrument to PC via a standard USB port with this cable. Testo™ USB Connection Cable connects Mini USB to Standard USB.
Brand:  Testo™ 0449 0047
Product Code. 10084333
  • 35.00€ / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • USB A to Mini B Connection
  • For use with Testo 330-1,330-2 LL and 335 data loggers
  • 6 foot USB connection cable
Specifications
Specifications
For Data Loggers: 330-1,330-2 LL and 335
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Testo™ USB Connection Cable > For Data Loggers: 330-1,330-2 LL and 335