Learn More
Thermo Scientific™ USA Standard Sieve, 60 Mesh
Brass frame and Stainless-steel mesh sieves
Brand:  Thermo Scientific™ 041200.ON
 View more versions of this product
Product Code. 11359557
  • 105.53€ / Each
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60 Mesh
1 in.
Sieve
3 in.
7.62cm
2.54cm
ASTM E11
Stainless Steel