missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Peek Low-Pressure Union Assembly
Make solid connections in low-pressure applications
Brand: Kinesis™ P-720
This item is not returnable.
View return policy
Description
Make solid connections in low-pressure applications
- Manufactured from inert polymers
- 1/4-28 flat-bottom ports
- Max pressure of 1000 psi (69 bar)
- 0.030 in. thru-hole diameter
- 0.41 μL swept volume
Specifications
Union | |
PEEK | |
Natural | |
MicroTight Sleeves; 2F-125/1P-553, 6-32 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction