Learn More
Zeiss™ Universal Mounting Frame
For use with mulitest dishes and petridishes in sizes 124 to 133mmL x 83 to 88mmW.
Brand: Zeiss™ 471717-0000-000
This item is not returnable.
View return policy
Description
Compatible Microscopes:
- Axio Observer 3
- Axio Observer 5
- Axio Observer 7
- Axio Observer.A1
- Axio Observer.D1
- Axio Observer.D1 TIRF 3
- Axio Observer.Z1
- Axio Observer.Z1 TIRF 3
- Axio Vert.A1
- Axio Vert.A1 FL
- Axio Vert.A1 FL-LED
- Axio Vert.A1 MAT
- Axiovert 200
- Axiovert 200 M
- Axiovert 25
- Axiovert 25 C
- Axiovert 25 CFL
- Axiovert 40 C
- Axiovert 40 CFL
- Primovert
Specifications
Universal Mounting Frame M-X | |
Multitest dishes with foot print 124 to 133mm x 83 to 88mm and Petri dishes dia.: 87 to 92mm | |
1 Ea. |