Learn More
Cytiva UNICORN™ start 1.1 Software on DVD
Product Code. 15873461 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
ÄKTA Start
Unit Size:
Each
Product Code. 15873461
Brand: Cytiva 29276964
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Conveniently control, monitor and evaluate protein purification. Cytiva UNICORN™ start 1.1 Software on DVD is the control software for the ÄKTA start, featuring intuitive design, easy reporting capabilities, real-time interactive process monitoring and robust data management.
UNICORN start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA start instrument, and to evaluate and share results. While ÄKTA start can function as a stand-alone system, UNICORN start software maximizes the performance of the instrument.
  • Simple and flexible method creation using predefined templates and phases (steps)
  • Intuitive graphic user interface enables users to perform many operations in a single-click
  • Enables real-time remote monitoring of the run and an intuitive way to control the run
  • Evaluate, compare, and share results; create and print PDF reports
  • Robust database storage with easy access to data, data security, and data integrity
TRUSTED_SUSTAINABILITY
Specifications
Type Version 1.1
Description System control software
For Use With (Equipment) ÄKTA Start
Includes DVD, activation code