Learn More
Qsonica Ultrasonic Homogenizer Converters
Qsonica Ultrasonic Homogenizer Converter works with either the Microson XL2000 or the Sonicator 3000 Ultrasonic Homogenizers to transform electrical signals into mechanical vibrations.
296.00€
Specifications
For Use With (Application) | Ideal for transform electrical signal into mechanical vibration |
---|
Produktcode | Marke | For Use With (Equipment) | Product Type | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | For Use With (Equipment) | Product Type | Preis | Menge & Verfügbarkeit | ||||
15142889
|
Qsonica
K4 |
Q500 and Q700 Sonicator™ | Converter Cable |
296.00€
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
||||
Beschreibung
- For use with Microson™ Model XL2000 or Sonicator™ 3000 Ultrasonic Homgenizers
- Transforms electrical signal into mechanical vibrations
Spezifikation
Ideal for transform electrical signal into mechanical vibration |