missing translation for 'onlineSavingsMsg'
Learn More

UK 14,304, Tocris Bioscience™

Product Code. 30000717
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α2 agonist

UK 14,304 is a full α2 adrenergic agonist. Centrally active following systemic administration in vivo.

Water-soluble Salt also available.

Chemical Identifiers

CAS 59803-98-4
Molecular Formula C11H10BrN5
Molecular Weight (g/mol) 292.14
MDL Number MFCD00153878
InChI Key XYLJNLCSTIOKRM-UHFFFAOYSA-N
Synonym brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
PubChem CID 2435
ChEBI CHEBI:3175
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
SMILES BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Specifications

Quantity 10 mg
Formula Weight 292.14
Percent Purity >98%
Chemical Name or Material UK 14, 304
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'