Learn More
SI Analytics™ Ubbelohde Viscometers for Manual Measurements
SI Analytics™ Ubbelohde Viscometers for Manual Measurements feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
Brand:  SI Analytics™ 285400029
 View more versions of this product
Product Code. 12008163
  • 505.00€ / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • The recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde Viscometer
Yes
0.005
Glass
Manual measurements
0.8 to 5 cSt
0.53 mm diameter
DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
290 mm
15 to 20 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products