missing translation for 'onlineSavingsMsg'
Learn More

(-)-U-50488 hydrochloride, Tocris Bioscience™

Product Code. 30001404
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard selective κ agonist. More active enantiomer of (±)-U-50488 (Cat. No. 0495)

More active enantiomer of (±)-U-50488 .

Racemate and (+)-enantiomer also available.

Chemical Identifiers

CAS 114528-79-9
Molecular Formula C19H29Cl3N2O2
Molecular Weight (g/mol) 423.80
MDL Number MFCD11044455
InChI Key UWPRPWMHQDPOFR-UHFFFAOYNA-N
Synonym --trans-1s,2s-u-50488 hydrochloride hydrate, trans-1s,2s-3,4-dichloro-n-methyl-n-2-1-pyrrolidinyl cyclohexyl benzeneacetamide hydrochloride hydrate, --trans-1s,2s-u-50488 hydrochloride hydrate, solid hplc, 2-3,4-dichlorophenyl-n-methyl-n-1s,2s-2-pyrrolidin-1-yl cyclohexyl acetamide hydrate hydrochloride
PubChem CID 16220065
IUPAC Name 2-(3,4-dichlorophenyl)-N-methyl-N-[2-(pyrrolidin-1-yl)cyclohexyl]acetamide hydrate hydrochloride
SMILES O.Cl.CN(C1CCCCC1N1CCCC1)C(=O)CC1=CC(Cl)=C(Cl)C=C1

Specifications

Quantity 10 mg
Formula Weight 405.79
Percent Purity >99%
Chemical Name or Material (-)-U-50488 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.