missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U 46619, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
PGH2 (TXA2) analog that is a potent and stable thromboxane A2 (TP) receptor agonist (EC50 = 0.035 μM). Potently stimulates TP receptor-mediated, but not other prostaglandin receptor-mediated responses in various in vitro preparations. Activates ERK-1 and ERK-2 in HEK 293 cells expressing TPα and TPβ receptors.
Specifications
Specifications
CAS | 56985-40-1 |
Molecular Formula | C21H34O4 |
Quantity | 1 mg |
Synonym | (5Z)-7-[(1R,4S,5S,6R)-6-[(1E,3S)-3-Hydroxy-1-octenyl]-2-oxabicyclo[2.2.1]hept-5-yl]-5-heptenoic acid |
InChI Key | LQANGKSBLPMBTJ-QQBDIJMNSA-N |
SMILES | CCCCCC(C=CC1C2CC(C1CC=CCCCC(=O)O)CO2)O |
IUPAC Name | (E)-7-[(1R,2S,3R,4R)-3-[(Z,3S)-3-hydroxyoct-1-enyl]-5-oxabicyclo[2.2.1]heptan-2-yl]hept-5-enoic acid |
Molecular Weight (g/mol) | 350.499 |
PubChem CID | 122173090 |
Formula Weight | 350.5 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'