missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Tyvek™ Apron

Product Code. 12017636
Click to view available options
110cm x 71cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Euro Protection™ Tyvek™ Apron provides limited chemical protection and is suitable for use in agribusiness and chemical industries.

  • Durable and resistant Tyvek™ construction
  • Superior quality
  • Limited chemical protection
  • Suitable for agribusiness, wet areas, chemical industry (against low-concentration inorganic chemicals)

Specifications

Material Tyvek
Color White
For Use With (Application) Agribusiness, wet areas, chemical industry
Dimensions (L x W) 110 cm x 71 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.