Learn More
Heidolph instruments Tygon™ (Standard) Tubing
For standard applications. For Single- and Multi-Channel use (not suitable with cassette small).
Brand: Heidolph instruments 525-69000-00
This item is not returnable.
View return policy
Description
- Non-toxic, non-oxidizing.
- Good resistance to acids, bases and inorganic media
- Very low gas permeability, good performance life
Specifications
Autoclavable | |
Borosilicate Glass | |
2.5 mm | |
Continuous/short time 0.5/1.5 bar |
6.3 mm | |
11.3 mm | |
Non-sterile |