Learn More
LABPLAS Twirl'Em Sample Bags
Twirl'em sample bags are safe, flexible, contaminant-free and easy to use.
Brand:  LABPLAS EPR-7012-FS
 View more versions of this product
Product Code. 17970295
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Twirl'Em sampling bags
  • Safety tabs
  • Clear, with writing area for easy labeling
  • Closure with 1 round wire and 1 flat wire
  • Available in various sizes
Specifications
Specifications
Sample Bag
Clear with White Block
Sterile
Yes
178 x 305 mm
178 mm
305 mm
Not Autoclavable
Single-use
76.2 μm
3 mil.
7 in.
12 in.