Learn More
WTW™ Turbidity Standard
Turbidity standards AMCO Clear™ with the highest precision and long-term stability are N.I.S.T. traceable, lot certified and not hazardous to your health.
Brand: WTW™ 600543
456.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Long-term precision and stability
- No health hazards
- Easy to dispose of
- Adapted to the respective device optics in their particle composition
- Suitable for applications in the lower measuring range such as drinking water
Specifications
Turbidity Standard | |
Calibration standards | |
0.02, 10, 100,1750 NTU |
Turb 555 IR | |
Cleaning Tissue and Marking Rings |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
WTW™ Turbidity Standard > For Use With: Turb 555 IR