Learn More
WTW™ Turbidimeter Cuvette
For automatic detection of the photLab™ turbidimeters and spectral photometers
Brand:  WTW™ 600601
 View more versions of this product
Product Code. 10181481
  • 196.00€ / Pack of 3
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cuvette
Turb 550, Turb 550 IR, Turb 555 and Turb 555 (IR) Laboratory Turbidimeter
3 Pack