Learn More
WTW™ Turb™ Calibration Kits
Get optimum calibration results for your turbidimeters with the Turb™ calibration kits.
Brand:  WTW™ 600547
 View more versions of this product
Product Code. 11798414
  • 348.00€ / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes Cleaning tissue and indexing rings
  • For use with Turb® 355 T, 335 IR Portable turbidimeter
  • Turbidity of 0.02 - 10 - 1000NTU
  • Calibration set
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turb 355 T, 335 IR Portable Turbidimeter
Calibration set
Cleaning tissue and indexing rings
0.02, 10, 1000 NTU