Learn More
WTW™ Turb™ Calibration Kits
Get optimum calibration results for your turbidimeters with the Turb™ calibration kits.
Brand:  WTW™ 600541
322.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10033022
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Includes cleaning tissue and 4 indexing rings
  • For use with Turb&reg 350 IR Portable turbidimeter
  • Turbidity of 0.02 - 10 - 100 - 1000NTU
  • Calibration set
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turb 350 IR Portable Turbidimeter
Calibration kit
Cleaning tissue and 4 indexing rings
0.02, 10, 100, 1000 NTU