Learn More
WTW™ Turb™ Calibration Kit
Get accurate test results with the Turb™ Calibration Kit.
Brand:  WTW™ 600542
 View more versions of this product
Product Code. 10688532
  • 349.00€ / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibration kit
  • For use with Turb™ 550T and Turb 550IR Turbidimeters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration Kit
Turb 550T and Turb 550IR Turbidimeters