Learn More
WTW™ Turb™ Calibration Kit
Get accurate test results with the Turb™ Calibration Kit.
Brand:  WTW™ 600560
 View more versions of this product
Product Code. 10631794
  • 356.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  •  Calibration kit
  • For use with Turb® 550 and Turb 555 Turbidimeters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration Standard Kit
Turb 430 IR Portable Turbidimeter