Learn More
Kartell™ Hoses Selector
Kartell™ Hoses Selector provides the nominal outside and inside diameter of hoses, allowing one to determine grip for connectors.
Brand:  Kartell™ 0175912
Product Code. 12617795
  • 3.75€ / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Description
  • Hoses selector gives nominal O.D. and I.D. of hoses
  • Use to determine grip for connectors
  • Selector includes following inside diameter in millimeters
    • 1, 2, 2.5, 3, 4, 4.5, 5, 6, 7, 8, 9.5, 10, 11, 12, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 25
  • Selector includes following outside diameter in millimeters
    • 1.5, 2, 2.5, 3, 4, 5.5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, 16, 17, 18, 21
Specifications
Specifications
Tube Selector
165 mm
90 mm
ABS
Hoses
Green