Learn More
Heidolph instruments Tube PolyBlock
Accommodate specifically sized vessels at your StarFish magnetic stirrer workstation. Heidolph™ Tube PolyBlock, is one of up to 5 StarFish PolyBlock segments that can be mixed at a single stirrer workstation to create various arrays of vessel size and type.
Brand: Heidolph instruments 505-80024-00
This item is not returnable.
View return policy
Description
- Accommodates one specific tube size.
- One of up to 5 segments per StarFish array.
Specifications
PolyBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers | |
9 x 12 mm Tubes |