Learn More
Heidolph instruments Tube PolyBlock
Accommodate specifically sized vessels at your StarFish magnetic stirrer workstation. Heidolph™ Tube PolyBlock, is one of up to 5 StarFish PolyBlock segments that can be mixed at a single stirrer workstation to create various arrays of vessel size and type.
Brand:  Heidolph instruments 505-80022-00
 View more versions of this product
Product Code. 11761659
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Accommodates one specific tube size.
  • One of up to 5 segments per StarFish array.
Specifications
Specifications
PolyBlock
Starfish Workstation, MR-Hei Magnetic Stirrers
3 x 24 mm Tubes