missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubacin, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Selective inhibitor of HDAC6; inhibits the second deacetylase domain (DD2). Does not inhibit HDAC6 histone deacetylase activity; reversibly inhibits α-tubulin deacetylation. Increases α-tubulin acetylation levels with no effect on histone acetylation or cell cycle progression.
Specifications
Specifications
CAS | 1350555-93-9 |
Molecular Formula | C41H43N3O7S |
Quantity | 1 mg |
Synonym | tubacin, n-4-2r,4r,6s-4-4,5-diphenyl-2-oxazolyl thio methyl-6-4-hydroxymethyl phenyl-1,3-dioxan-2-yl phenyl-n'-hydroxyoctanediamide, n1-4-2r,4r,6s-4-4,5-diphenyloxazol-2-yl thio methyl-6-4-hydroxymethyl phenyl-1,3-dioxan-2-yl phenyl-n8-hydroxyoctanediamide, d04kqf, tubacin bml-gr362, octanediamide, n1-4-2r,4r,6s-4-4,5-diphenyl-2-oxazolyl thio methyl-6-4-hydroxymethyl phenyl-1,3-dioxan-2-yl phenyl-n8-hydroxy-, rel, tubacin hplc, unii-02c2g1d30d component, n-4-2r,4r,6s-4-4,5-diphenyl-1,3-oxazol-2-yl sulfanyl methyl-6-4-hydroxymethyl phenyl-1,3-dioxan-2-yl phenyl-n-hydroxyoctanediamide, n-4-2r,4r,6s-4-4,5-diphenyl-1,3-oxazol-2-yl sulfanylmethyl-6-4-hydroxymethyl phenyl-1,3-dioxan-2-yl phenyl-n'-hydroxyoctanediamide |
InChI Key | BHUZLJOUHMBZQY-YXQOSMAKSA-N |
SMILES | C1C(OC(OC1C2=CC=C(C=C2)CO)C3=CC=C(C=C3)NC(=O)CCCCCCC(=O)NO)CSC4=NC(=C(O4)C5=CC=CC=C5)C6=CC=CC=C6 |
IUPAC Name | N-[4-[(2R,4R,6S)-4-[(4,5-diphenyl-1,3-oxazol-2-yl)sulfanylmethyl]-6-[4-(hydroxymethyl)phenyl]-1,3-dioxan-2-yl]phenyl]-N'-hydroxyoctanediamide |
Molecular Weight (g/mol) | 721.869 |
PubChem CID | 6675804 |
Formula Weight | 721.86 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'