missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Refrigerators - Boxed
GREENER_CHOICE

The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

Brand:  Thermo Scientific™ TSG1500REGBV

 View more versions of this product

Product Code. 17127738

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from 400 L to 1500 L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG Series General Purpose Laboratory Refrigerator
+5°C
Glass
8 Adjustable
Steel
Painted Steel
Yes
1 Rear Access Port
Adaptive Defrost
Alarm
Smart-Vue Pro Compatible
837 x 1440 x 2082 mm
230 V
50 Hz
283 kg
1500 L
R290
2
30 kg
Painted Steel
CE
Digital Microprocessor
36 Months
Boxed
Key Lock
690 x 1300 x 1510 mm
EU
230 V/50 Hz
Energy Efficient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions