missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Refrigerators - Crated
GREENER_CHOICE

The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

Brand:  Thermo Scientific™ TSG700RESCV

 View more versions of this product

Product Code. 17127768

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from 400L to 1500L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG Series General Purpose Laboratory Refrigerator
2°C to 10°C
R290
1
30 kg
Painted Steel
Yes
1 Rear Access Port
Adaptive Defrost
Alarm
Smart-Vue Pro Compatible
840 x 750 x 2100 mm
230 V
Energy Efficient
700 L
+5°C
Solid
4 Adjustable
Painted Steel
CE
Digital Microprocessor
12 Months
Crated
Key Lock
686 x 600 x 1505 mm
EU
50 Hz
165 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions