missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Freezers - Boxed
GREENER_CHOICE

The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

Brand:  Thermo Scientific™ TSG1500FESBV

5864.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 17187728

  • 7477.20€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from solid door 400 L to 1500 L double door.
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG Series General Purpose Laboratory Freezer
-12°C to -30°C
Solid
Steel
8 Adjustable
CE
Touchscreen Control with Password Protection
36 Months
Boxed
837 x 1440 x 2082 mm
Smart-Vue Pro Compatible
230 V
50 Hz
263 kg
1500 L
-20°C
2
Painted Steel
30 kg
Yes
1 Rear Access Port
Adaptive Defrost
690 x 1300 x 1510 mm
Alarm
EU
230V/50Hz
R290
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.