missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, Serva Electrophoresis™
Click to view available options
Quantity:
1 kg
Specifications
Specifications
CAS | 77-86-1 |
Melting Point | 170°C |
Color | White |
Density | 1.35g/cm3 |
pH | 10 to 11.5 |
Molecular Formula | C4H11NO3 |
Quantity | 1 kg |
Synonym | trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, trizma, 2-amino-2-hydroxymethyl-1,3-propanediol, 2-amino-2-hydroxymethyl propane-1,3-diol, tris buffer |
Solubility Information | 800g/L |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Show More |
Safety and Handling
Product Identifier
- Tris(hydroxymethyl)aminomethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'