Learn More
Fisherbrand™ Triple Instrument Bundle
Whether you need to spin-down, shake, or stir, this Triple Instrument Bundle has you covered.
Brand:  Fisherbrand™ 120950
140.89 EUR valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Product Code. 17862966
  • 1050.00€ / 1 set
Edge
Explore more special offers
Description
Description
  • Get the full functionality of three machines in two pieces of equipment! Complete with a Fisherbrand Silicone Lab Mat to protect your workspace from spills and wear.
  • Fisherbrand Centrifuge/Stirrer: The industry's first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 L), combined into one unit. Made durable and chemically resistant with easy cleaning and maintenance-free operation. Its compact and low-profile design optimizes limited bench space and minimizes clutter in the lab.
  • Fisherbrand Mini Vortexer: Small footprint Vortexer offering maximum vortex shaking for a variety of tube sizes. A simple touch start operation allows for immediate sample mixing with one hand and automatic stop when tube is lifted.
  • Fisherbrand Silicone Lab Mat: This FDA food-grade silicone labmat is suitable for various laboratory settings, including cold rooms, clean rooms, and sterile suites, where high sterility is essential. Effectively safeguards countertops from temperatures up to 200°C (392°F) and demonstrates general chemical resistance to moderate oxidizing chemicals.
Specifications
Specifications
100/240 V
Promotions
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095
    Expires2025-04-15
    Days
    15
    Hours
    11
    Minutes
    51
    Seconds
    48
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
    New Offer
    Save Now
Product Content Correction
Product Title
Fisherbrand™ Triple Instrument Bundle > Fisherbrand Triple Instrument