missing translation for 'onlineSavingsMsg'
Learn More

Triphenylethylene, 98+%, Thermo Scientific Chemicals

Product Code. 11420783
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-72-0
Molecular Formula C20H16
Molecular Weight (g/mol) 256.348
MDL Number MFCD00004765
InChI Key MKYQPGPNVYRMHI-UHFFFAOYSA-N
Synonym triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b
PubChem CID 6025
ChEBI CHEBI:35034
IUPAC Name 1,2-diphenylethenylbenzene
SMILES C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 67°C to 71°C
Boiling Point 219°C to 221°C (14 mmHg)
Quantity 10 g
Beilstein 1867462
Formula Weight 256.35
Percent Purity ≥98%
Chemical Name or Material Triphenylethylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'